Merck
  • Computational design of virus-like protein assemblies on carbon nanotube surfaces.
Computational design of virus-like protein assemblies on carbon nanotube surfaces.
Science (New York, N.Y.) (2011-05-28)
Gevorg Grigoryan, Yong Ho Kim, Rudresh Acharya, Kevin Axelrod, Rishabh M Jain, Lauren Willis, Marija Drndic, James M Kikkawa, William F DeGrado
ABSTRACT
There is a general need for the engineering of protein-like molecules that organize into geometrically specific superstructures on molecular surfaces, directing further functionalization to create richly textured, multilayered assemblies. Here we describe a computational approach whereby the surface properties and symmetry of a targeted surface define the sequence and superstructure of surface-organizing peptides. Computational design proceeds in a series of steps that encode both surface recognition and favorable intersubunit packing interactions. This procedure is exemplified in the design of peptides that assemble into a tubular structure surrounding single-walled carbon nanotubes (SWNTs). The geometrically defined, virus-like coating created by these peptides converts the smooth surfaces of SWNTs into highly textured assemblies with long-scale order, capable of directing the assembly of gold nanoparticles into helical arrays along the SWNT axis.