Merck
Defining the CD59-C9 binding interaction.
The Journal of biological chemistry (2006-07-18)
Yuxiang Huang, Fei Qiao, Ruben Abagyan, Starr Hazard, Stephen Tomlinson
ABSTRACT
CD59 is a membrane glycoprotein that regulates formation of the cytolytic membrane attack complex (MAC or C5b-9) on host cell membranes. It functions by binding to C8 (alpha chain) and C9 after their structural rearrangement during MAC assembly. Previous studies indicated that the CD59 binding site in C9 was located within a 25-residue disulfide-bonded loop, and in C8alpha was located within a 51-residue sequence that overlaps the CD59 binding region of C9. By peptide screens and the use of peptides in binding assays, functional assays, and computer modeling and docking studies, we have identified a 6-residue sequence of human C9, spanning residues 365-371, as the primary CD59 recognition domain involved in CD59-mediated regulation of MAC formation. The data also indicate that both C8alpha and C9 bind to a similar or overlapping site on CD59. Furthermore, data from CD59-peptide docking models are consistent with the C9 binding site on CD59 located at a hydrophobic pocket, putatively identified previously by CD59 mutational and modeling studies.