Merck
  • Homogeneous Escherichia coli FPG protein. A DNA glycosylase which excises imidazole ring-opened purines and nicks DNA at apurinic/apyrimidinic sites.
Homogeneous Escherichia coli FPG protein. A DNA glycosylase which excises imidazole ring-opened purines and nicks DNA at apurinic/apyrimidinic sites.
The Journal of biological chemistry (1990-03-05)
S Boiteux, T R O'Connor, F Lederer, A Gouyette, J Laval
ABSTRACT
The repair of 2,6-diamino-4-hydroxy-5-N-methyl-formamidopyrimidine (Fapy) residues in DNA is performed by a Fapy-DNA glycosylase activity which is encoded for by the fpg gene in Escherichia coli. Besides DNA glycosylase activity, this protein, the FPG protein, is endowed with an EDTA-resistant activity nicking DNA at apurinic/apyrimidinic (AP) sites. To overproduce the FPG protein, the fpg gene was placed under the control of the tac promoter in the expression vector pKK223-3 yielding the pFPG230 plasmid. The production of the FPG protein in cells harboring the pFPG230 plasmid was 800-fold higher than that of the wild type strain after induction by isopropyl-beta-D-thio-galactopyranoside. From these cells, the FPG protein was purified to homogeneity in sufficient quantity to study its physical and catalytic properties. In its active form, the FPG protein is a globular monomer of 31 kDa and has an experimentally measured isoelectric point of 8.5. When the FPG protein is heat-denatured in the presence of EDTA the two activities are more rapidly inactivated than when heated in the absence of EDTA, suggesting that the FPG protein possesses a tightly bound metal ion. Atomic absorption spectrophotometric analysis shows that there is one zinc/FPG protein molecule. The FPG protein is different from previously described DNA glycosylases and AP-nicking enzymes in E. coli. The contribution of the AP-nicking activity associated with the FPG protein represents 10-20% of the total EDTA-resistant AP-nicking activities in E. coli.