Merck
  • Purification of active TFIID from Saccharomyces cerevisiae. Extensive promoter contacts and co-activator function.
Purification of active TFIID from Saccharomyces cerevisiae. Extensive promoter contacts and co-activator function.
The Journal of biological chemistry (2004-09-28)
Roy Auty, Hanno Steen, Lawrence C Myers, Jim Persinger, Blaine Bartholomew, Steven P Gygi, Stephen Buratowski
ABSTRACT
The basal transcription factor TFIID is composed of the TATA-binding protein (TBP) and 14 TBP-associated factors (TAFs). Although TBP alone binds to the TATA box of DNA and supports basal transcription, the TAFs have essential functions that remain poorly defined. In order to study its properties, TFIID was purified from Saccharomyces cerevisiae using a newly developed affinity tag. Analysis of the final elution by mass spectrometry confirms the presence of all the known TAFs and TBP, as well as Rsp5, Bul1, Ubp3, Bre5, Cka1, and Cka2. Both Taf1 and Taf5 are ubiquitinated, and the ubiquitination pattern of TFIID changes when BUL1 or BRE5 is deleted. Purified TFIID binds specifically to promoter DNA in a manner stabilized by TFIIA, and these complexes can be analyzed by native gel electrophoresis. Phenanthroline-copper footprinting and photoaffinity cross-linking indicate that TFIID makes extensive contacts upstream and downstream of the TATA box. TFIID supports basal transcription and activated transcription, both of which are enhanced by TFIIA.