Merck
  • Immunohistochemical study of nasopharyngeal carcinoma using monoclonal keratin antibodies.
Immunohistochemical study of nasopharyngeal carcinoma using monoclonal keratin antibodies.
The American journal of pathology (1984-10-01)
S R Shi, M L Goodman, A K Bhan, B Z Pilch, L B Chen, T T Sun
ABSTRACT
Nasopharyngeal carcinoma (NPC) provides a unique opportunity to evaluate distinctive epidemiologic features and a possible etiologic relationship with Epstein-Barr virus (EBV) in human malignancy. The lack of a uniformly accepted pathologic classification for NPC has limited the application of this data, although the World Health Organization (WHO) developed a classification that may solve this problem. Monoclonal keratin antibodies were used for staining of NPC for evaluation of its assistance in diagnosis and classification. In the present immunohistochemical study, monoclonal keratin antibodies, designated AE1, AE2, and AE3, and a polyclonal keratin antibody (RAK) were used for study of the presence of keratin in 121 cases of NPC obtained from China and the United States. AE1 monoclonal antibody, which recognizes keratin protein classes 56.5K, 50K, and 40K, was shown to be the most sensitive and specific for NPC tumor cells among the keratin antibodies studied. In addition, some different keratin expression patterns could be identified between different kinds of epithelium and different tumor groups, with possible relevance to the histogenesis of the histologic subtypes of NPC.
Sigma-Aldrich
Anti-Keratin Epithelial Antibody, clone AE3, clone AE3, Chemicon®, from mouse