Merck
  • N(G)-nitro-L-arginine impairs the anticonvulsive action of ethosuximide against pentylenetetrazol.
N(G)-nitro-L-arginine impairs the anticonvulsive action of ethosuximide against pentylenetetrazol.
European journal of pharmacology (1999-03-19)
S J Czuczwar, P Tutka, P Klonowski, Z Kleinrok
ABSTRACT
N(G)-nitro-L-arginine (NNA; an inhibitor of nitric oxide synthase) in a dose of 40 mg/kg impaired the protective activity of ethosuximide against the clonic phase of pentylenetetrazol-induced seizures in mice. The ED50 value of ethosuximide was significantly increased from 108 to 158 mg/kg. NNA (40 mg/kg) was ineffective against the protective effects of diazepam, phenobarbital and valproate against pentylenetetrazol-induced seizures. NNA (40 mg/kg) did not influence the plasma levels of the antiepileptic drugs studied, so a pharmacokinetic interaction is not probable. L-Arginine (500 mg/kg) prevented the NNA-induced reduction of the anticonvulsive activity of ethosuximide. It can be concluded that nitric oxide participates in the expression of the anticonvulsive action of ethosuximide, but not that of diazepam, phenobarbital and valproate, against pentylenetetrazol-induced seizures.