Merck
  • Investigations on the acute effects of neuropeptides on the pituitary-adrenocortical function in normal and cold-stressed rats. II. Neurotensin and neuromedin N.
Investigations on the acute effects of neuropeptides on the pituitary-adrenocortical function in normal and cold-stressed rats. II. Neurotensin and neuromedin N.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie (1995-01-01)
L K Malendowicz, G G Nussdorfer, A Markowska, C Tortorella, M Nowak
ABSTRACT
The effects of a subcutaneous bolus injection of 2 micrograms neurotensin (NT) or neuromedin N (NMN) on the function of the hypothalamo-pituitary-adrenocortical axis were investigated in both normal and cold-stressed rats. The blood concentrations of ACTH, corticosterone (B) and aldosterone (ALDO) were measured by specific radioimmunoassays 1, 2 or 4 h after the neuropeptide administration. Cold stress enhanced plasma levels of ACTH, B and ALDO, and these rises lasted unchanged until 4 h. NT did not affect either basal or stress-stimulated plasma levels of ACTH and B, while it lowered the plasma ALDO concentration at 4 h in normal rats and increased it at 1 h in stressed animals. NMN did not change the basal plasma level of ACTH, but it did markedly raise blood levels of both B and ALDO; on the other hand, in cold-stressed rats NMN strongly depressed ACTH response and decreased B plasma concentration at 2 h, without evoking apparent changes in ALDO response. In light of these findings the following conclusions and hypotheses can be drawn and suggested: (i) NT and NMN, when administered at a relatively high dose, do not affect ACTH release in rats under basal conditions; (ii) NMN, but not NT, is able to prevent cold stress-induced stimulation of ACTH secretion, probably by inhibiting hypothalamic thermoregulatory centers; and (iii) NT and NMN exert direct adrenocortical antisecretagogue and secretagogue effects, respectively, which could explain the evident lack of correlation between the levels of circulating ACTH and the plasma concentrations of the main adrenal steroid hormones in both normal and stressed rats after neuropeptide administration.