Merck
  • Induction of synthesis of tetrahydropyrimidine derivatives in Streptomyces strains and their effect on Escherichia coli in response to osmotic and heat stress.
Induction of synthesis of tetrahydropyrimidine derivatives in Streptomyces strains and their effect on Escherichia coli in response to osmotic and heat stress.
Journal of bacteriology (1996-01-01)
G Malin, A Lapidot
ABSTRACT
The metabolic responses of a number of Streptomyces strains to osmotic and heat stress were studied by 13C nuclear magnetic resonance spectroscopy. During cell growth in a chemically defined medium supplemented with 0.5 M NaCl, tetrahydropyrimidine derivatives (THPs), 2-methyl-4-carboxy-5-hydroxy-3,4,5,6-tetrahydropyrimidine [THP(A)] and, to a lesser extent, 2-methyl-4-carboxy-3,4,5,6-tetrahydropyrimidine [THP(B)], were found to accumulate in a significant amount in all bacteria examined. In addition, when the growth temperature was shifted from 30 to 39 degrees C, the intracellular concentration of THP(A) increased significantly. Moreover, exogenously provided THP(A) or THP(B) or both reversed inhibition of Escherichia coli growth caused by osmotic stress and increased temperature. Although the ability of Streptomyces strains to tolerate high concentrations of NaCl is well known, very little is known about the osmoregulatory strategy in Streptomyces strains. Similarly, the mechanism by which compatible solutes accumulate in a variety of microorganisms is not understood. Our findings suggest the possibility of a novel mechanism of protection of DNA against salt and heat stresses involving the THPs.