- Influence of the addition of Amaranthus mantegazzianus flour on the nutritional and health properties of pasta
Influence of the addition of Amaranthus mantegazzianus flour on the nutritional and health properties of pasta
Cogent Food & Agriculture (2016)
Martinez CS, et al.