Merck
What Do You Get from DNA-Encoded Libraries?
ACS medicinal chemistry letters (2018-05-26)
Alexander L Satz
ABSTRACT
Use of DNA-encoded libraries (DELs) in the pharmaceutical industry has rapidly increased. We discuss what to expect when you run a DEL screen and contemplate guidelines for library design. Additionally, we consider some visionary work and extrapolate to the future.