Merck
MicroRNA-101 inhibits angiogenesis via COX-2 in endometrial carcinoma.
Molecular and cellular biochemistry (2018-02-07)
Ying Liu, Haiyan Li, Congying Zhao, Hanbing Jia
ABSTRACT
Abnormal angiogenesis is critically involved in tumor progression and metastasis including endometrial cancer and is regulated by microRNAs such as microRNA-101 (miR-101). We hypothesize that miR-101 expression is disrupted in endometrial cancer and modulation of miR-101 levels is sufficient to regulate tumor growth through angiogenesis. We examined the expression levels of miR-101 and factors involved in angiogenesis in the patients with endometrial cancer. We also overexpressed or inhibited miR-101 in RL-95-2 cells and examined their effects on cell toxicity and tumor growth. Finally, we determined if miR-101 regulated tumorigenesis through cyclooxygenase-2 (COX-2). We found that miR-101 levels were significantly reduced. Factors involved in angiogenesis included vascular endothelial growth factor-A (VEGF-A), thrombospondin-1 (TSP-1), cyclooxygenase-2 (COX-2), prostaglandin E2 (PGE2), and aromatase (P450arom), which were increased in endometrial carcinoma. Modulation of miR-101 level was sufficient to affect tumor growth. Finally, we found that the effects of miR-101 inhibition on tumor growth were suppressed by COX-2 inhibition. Our results suggest that modulating miR-101 and COX-2 levels or their activity may be a potential therapeutic strategy for endometrial cancer.