Merck
  • Expression Profile of LGR5 and Its Prognostic Significance in Colorectal Cancer Progression.
Expression Profile of LGR5 and Its Prognostic Significance in Colorectal Cancer Progression.
The American journal of pathology (2018-07-24)
Bo Gun Jang, Hye Sung Kim, Weon Young Chang, Jeong Mo Bae, Woo Ho Kim, Gyeong Hoon Kang
ABSTRACT
We investigated the expression profile of leucine-rich, repeat-containing, G-protein-coupled receptor 5 (LGR5) during colorectal cancer (CRC) progression and determined the prognostic impact of LGR5 in a large cohort of CRC samples. LGR5 expression was higher in CRCs than in normal mucosa, and was not associated with other cancer stem cell markers. LGR5 positivity was observed in 68% of 788 CRCs and was positively correlated with older age, moderately to well-differentiated cells, and nuclear β-catenin expression. Enhanced LGR5 expression remained persistent during the adenoma-carcinoma transition, but markedly declined in the budding cancer cells at the invasive fronts, which was not due to altered wingless-type mouse mammary tumor virus integration site family (Wnt) or epithelial-mesenchymal transition signaling. LGR5 showed negative correlations with microsatellite instability and CpG island methylator phenotype, and was not associated with KRAS or BRAF mutation. Notably, LGR5 positivity was an independent prognostic marker for better clinical outcomes in CRC patients. LGR5 overexpression attenuated tumor growth by decreasing ERK phosphorylation along with decreased colony formation and migration abilities in DLD1 cells. Likewise, knockdown of LGR5 expression resulted in a decline in the colony-forming and migration capacities in LoVo cells. Taken together, our data suggest a suppressive role of LGR5 in CRC progression.