Merck
  • Lymphocyte activation gene 3 (Lag3) expression is increased in prion infections but does not modify disease progression.
Lymphocyte activation gene 3 (Lag3) expression is increased in prion infections but does not modify disease progression.
Scientific reports (2018-10-04)
Yingjun Liu, Silvia Sorce, Mario Nuvolone, Julie Domange, Adriano Aguzzi
ABSTRACT
Prion diseases, Alzheimer's disease and Parkinson's disease (PD) are fatal degenerative disorders that share common neuropathological and biochemical features, including the aggregation of pathological protein conformers. Lymphocyte activation gene 3 (Lag3, also known as CD223) is a member of the immunoglobulin superfamily of receptors expressed on peripheral immune cells, microglia and neurons, which serves as a receptor for α-synuclein aggregates in PD. Here we examined the possible role of Lag3 in the pathogenesis of prion diseases. Through quantitative real-time PCR and RNA-sequencing, we found that the expression levels of Lag3 were relatively low in the adult mouse brains, yet its expression was increased after prion infection. However, we failed finding significant differences regarding the incubation time, PrPSc load, neurodegeneration, astrocyte and microglia reactions and inflammatory gene expression between the Lag3 knockout mice and wild-type littermate controls after prion infection. We conclude that loss of Lag3 has no significant influence on prion disease pathogenesis. Considering that Lag3 is an immune checkpoint receptor, our results suggest that immune checkpoint inhibition (an increasingly prevalent therapeutic modality against many types of cancer) might not exert positive or negative effects on the progression of prion diseases.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse