Merck
  • Immune-suppression by OsHV-1 viral infection causes fatal bacteraemia in Pacific oysters.
Immune-suppression by OsHV-1 viral infection causes fatal bacteraemia in Pacific oysters.
Nature communications (2018-10-13)
Julien de Lorgeril, Aude Lucasson, Bruno Petton, Eve Toulza, Caroline Montagnani, Camille Clerissi, Jeremie Vidal-Dupiol, Cristian Chaparro, Richard Galinier, Jean-Michel Escoubas, Philippe Haffner, Lionel Dégremont, Guillaume M Charrière, Maxime Lafont, Abigaïl Delort, Agnès Vergnes, Marlène Chiarello, Nicole Faury, Tristan Rubio, Marc A Leroy, Adeline Pérignon, Denis Régler, Benjamin Morga, Marianne Alunno-Bruscia, Pierre Boudry, Frédérique Le Roux, Delphine Destoumieux-Garzόn, Yannick Gueguen, Guillaume Mitta
ABSTRACT
Infectious diseases are mostly explored using reductionist approaches despite repeated evidence showing them to be strongly influenced by numerous interacting host and environmental factors. Many diseases with a complex aetiology therefore remain misunderstood. By developing a holistic approach to tackle the complexity of interactions, we decipher the complex intra-host interactions underlying Pacific oyster mortality syndrome affecting juveniles of Crassostrea gigas, the main oyster species exploited worldwide. Using experimental infections reproducing the natural route of infection and combining thorough molecular analyses of oyster families with contrasted susceptibilities, we demonstrate that the disease is caused by multiple infection with an initial and necessary step of infection of oyster haemocytes by the Ostreid herpesvirus OsHV-1 µVar. Viral replication leads to the host entering an immune-compromised state, evolving towards subsequent bacteraemia by opportunistic bacteria. We propose the application of our integrative approach to decipher other multifactorial diseases that affect non-model species worldwide.