Merck
  • P-cadherin is coexpressed with CD44 and CD49f and mediates stem cell properties in basal-like breast cancer.
P-cadherin is coexpressed with CD44 and CD49f and mediates stem cell properties in basal-like breast cancer.
Stem cells (Dayton, Ohio) (2012-03-06)
André Filipe Vieira, Sara Ricardo, Matthew Paul Ablett, Maria Rita Dionísio, Nuno Mendes, André Albergaria, Gillian Farnie, Renê Gerhard, Jorge F Cameselle-Teijeiro, Raquel Seruca, Fernando Schmitt, Robert B Clarke, Joana Paredes
ABSTRACT
Although the luminal progenitor cell of the normal mammary gland hierarchy has been proposed as the cell-of-origin for basal-like breast cancers, finding the cancer stem cell (CSC) phenotype for this malignancy has proven a difficult task, mostly due to the lack of specific markers. Recently, basal-like sporadic and familial cases of breast cancer have been linked to BRCA1 gene inactivation, which enables the upregulation of the target-repressed CDH3/P-cadherin gene, an important biomarker of basal-like breast carcinomas. Previously, we demonstrated that P-cadherin overexpression can mediate aggressive behavior in these tumors. Thus, our aim was to test whether P-cadherin mediates stem cell properties in basal-like breast carcinomas. Using a series of breast cancer cell lines and primary tumors, we showed that P-cadherin was directly associated with the expression of the breast stem markers CD44, CD49f, and aldehyde dehydrogenase 1 in the basal subtype. Moreover, cell population enriched for P-cadherin expression comprised increased in vitro mammosphere-forming efficiency and capacity to grow colonies in three-dimensional cultures as well as greater tumorigenicity. Importantly, an association was found with stem-/progenitor-like phenotypes of the breast, including the luminal progenitor population, CD49f(+) CD24(+). Additionally, P-cadherin expression conferred resistance to x-ray-induced cell death, sustaining a role for this molecule in another stem cell property. In summary, we demonstrated, for the first time, that P-cadherin mediates stem cell properties, which could be explored in order to better define the CSC phenotype of basal-like breast tumors and the cell-of-origin of this malignancy.