Merck
  • LIN28 phosphorylation by MAPK/ERK couples signalling to the post-transcriptional control of pluripotency.
LIN28 phosphorylation by MAPK/ERK couples signalling to the post-transcriptional control of pluripotency.
Nature cell biology (2016-12-20)
Kaloyan M Tsanov, Daniel S Pearson, Zhaoting Wu, Areum Han, Robinson Triboulet, Marc T Seligson, John T Powers, Jihan K Osborne, Susan Kane, Steven P Gygi, Richard I Gregory, George Q Daley
ABSTRACT
Signalling and post-transcriptional gene control are both critical for the regulation of pluripotency, yet how they are integrated to influence cell identity remains poorly understood. LIN28 (also known as LIN28A), a highly conserved RNA-binding protein, has emerged as a central post-transcriptional regulator of cell fate through blockade of let-7 microRNA biogenesis and direct modulation of mRNA translation. Here we show that LIN28 is phosphorylated by MAPK/ERK in pluripotent stem cells, which increases its levels via post-translational stabilization. LIN28 phosphorylation had little impact on let-7 but enhanced the effect of LIN28 on its direct mRNA targets, revealing a mechanism that uncouples LIN28's let-7-dependent and -independent activities. We have linked this mechanism to the induction of pluripotency by somatic cell reprogramming and the transition from naive to primed pluripotency. Collectively, our findings indicate that MAPK/ERK directly impacts LIN28, defining an axis that connects signalling, post-transcriptional gene control, and cell fate regulation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)