Merck
An interweaving MOF with high hydrogen uptake.
Journal of the American Chemical Society (2006-03-23)
Daofeng Sun, Shengqian Ma, Yanxiong Ke, David J Collins, Hong-Cai Zhou
ABSTRACT
An interweaving copper metal-organic framework possessing high porosity and high hydrogen uptake has been successfully designed and synthesized.