Merck
  • Targetted inhibition of CD74 attenuates adipose COX-2-MIF-mediated M1 macrophage polarization and retards obesity-related adipose tissue inflammation and insulin resistance.
Targetted inhibition of CD74 attenuates adipose COX-2-MIF-mediated M1 macrophage polarization and retards obesity-related adipose tissue inflammation and insulin resistance.
Clinical science (London, England : 1979) (2018-05-19)
Pei-Chi Chan, Ting-Ni Wu, Ying-Chuan Chen, Chieh-Hua Lu, Martin Wabitsch, Yu-Feng Tian, Po-Shiuan Hsieh
ABSTRACT
Adipose tissue (AT) inflammation is crucial to the development of obesity-associated insulin resistance. Our aim was to investigate the contribution of cyclooxygenase-2 (COX-2)/macrophage migration inhibitory factor (MIF)-mediated cross-talk between hypertrophic adipocytes and macrophages to the etiology of AT inflammation and the involvement of CD74 using human SGBS adipocytes, THP-1 macrophages and mice fed a high-fat (HF) diet. The MIF and CD74 mRNA levels in the adipocytes and stromal vascular cells (SVCs) of white fat were highly correlated with body weight (BW), homeostatic model assessment for insulin resistance (HOMA-IR), and adipose macrophage marker expression levels, especially those in SVCs. COX-2 inhibition suppressed the elevation of MIF production in HF white adipocytes as well as palmitate and hypoxic-treated SGBS adipocytes. Treatment of adipocytes transfected with shCOX-2 and siMIF or subjected to MIF depletion in the medium reversed the pro-inflammatory responses in co-incubated THP-1 cells. Inhibition of NF-κB activation reversed the COX2-dependent MIF secretion from treated adipocytes. The targetted inhibition of macrophage CD74 prevented M1 macrophage polarization in the above co-culture model. The COX-2-dependent increases in CD74 gene expression and MIF release in M1-polarized macrophages facilitated the expression of COX-2 and MIF in co-cultured SGBS adipocytes. CD74 shRNA intravenous injection suppressed HF-induced AT M1 macrophage polarization and inflammation as well as insulin resistance in mice. The present study suggested that COX-2-mediated MIF secretion through NF-κB activation from hypertrophic and hypoxic adipocytes as well as M1 macrophages might substantially contribute to the phenotypic switch of AT macrophages through CD74 in obesity. Inhibition of CD74 could attenuate AT inflammation and insulin resistance in the development of HF diet-induced obesity.