Merck
  • The C-terminus of PRK2/PKNgamma is required for optimal activation by RhoA in a GTP-dependent manner.
The C-terminus of PRK2/PKNgamma is required for optimal activation by RhoA in a GTP-dependent manner.
Archives of biochemistry and biophysics (2008-10-07)
Wee Guan Lim, Xiao Chen, Jun-Ping Liu, Bee Jen Tan, Shufeng Zhou, Adam Smith, Nathaniel Lees, Liansheng Hou, Fukang Gu, Xi Yong Yu, Yaomin Du, Derek Smith, Chandra Verma, Ke Liu, Wei Duan
ABSTRACT
PRK2/PKNgamma is a Rho effector and a member of the protein kinase C superfamily of serine/threonine kinases. Here, we explore the structure-function relationship between various motifs in the C-terminal half of PRK2 and its kinase activity and regulation. We report that two threonine residues at conserved phosphoacceptor position in the activation loop and the turn motif are essential for the catalytic activity of PRK2, but the phosphomimetic Asp-978 at hydrophobic motif is dispensable for kinase catalytic competence. Moreover, the PRK2-Delta958 mutant with the turn motif truncated still interacts with 3-phosphoinositide-dependent kinase-1 (PDK-1). Thus, both the intact hydrophobic motif and the turn motif in PRK2 are dispensable for the binding of PDK-1. We also found that while the last seven amino acid residues at the C-terminus of PRK2 are not required for the activation of the kinase by RhoA in vitro, however, the extreme C-terminal segment is critical for the full activation of PRK2 by RhoA in cells in a GTP-dependent manner. Our data suggest that the extreme C-terminus of PRK2 may represent a potential drug target for effector-specific pharmacological intervention of Rho-medicated biological processes.