- Insulin-dependent stimulation of protein synthesis by phosphorylation of a regulator of 5'-cap function.
Insulin-dependent stimulation of protein synthesis by phosphorylation of a regulator of 5'-cap function.
Nature (1994-10-27)
A Pause, G J Belsham, A C Gingras, O Donzé, T A Lin, J C Lawrence, N Sonenberg
PMID7935836
ABSTRACT
The cloning is described of two related human complementary DNAs encoding polypeptides that interact specifically with the translation initiation factor eIF-4E, which binds to the messenger RNA 5'-cap structure. Interaction of these proteins with eIF-4E inhibits translation but treatment of cells with insulin causes one of them to become hyperphosphorylated and dissociate from eIF-4E, thereby relieving the translational inhibition. The action of this new regulator of protein synthesis is therefore modulated by insulin, which acts to stimulate the overall rate of translation and promote cell growth.