Merck
  • Synthesis of (nor)tropeine (di)esters and allosteric modulation of glycine receptor binding.
Synthesis of (nor)tropeine (di)esters and allosteric modulation of glycine receptor binding.
Bioorganic & medicinal chemistry (2007-12-07)
Gábor Maksay, Péter Nemes, Zoltán Vincze, Timea Bíró
ABSTRACT
(Hetero)aromatic mono- and diesters of tropine and nortropine were prepared. Modulation of [3H]strychnine binding to glycine receptors of rat spinal cord was examined with a ternary allosteric model. The esters displaced [3H]strychnine binding with nano- or micromolar potencies and strong negative cooperativity. Coplanarity and distance of the ester moieties of diesters affected the binding affinity being nanomolar for isophthaloyl-bistropane and nortropeines. Nortropisetron had the highest affinity (K(A) approximately 10 nM). Two esters displayed negative cooperativity with glycine in displacement, while three esters of low-affinity and nortropisetron exerted positive cooperativity with glycine.