- PhenoFluor: Practical Synthesis, New Formulation, and Deoxyfluorination of Heteroaromatics.
PhenoFluor: Practical Synthesis, New Formulation, and Deoxyfluorination of Heteroaromatics.
Organic process research & development (2014-09-02)
Teppei Fujimoto, Fabian Becker, Tobias Ritter
PMID25177149
ABSTRACT
We report a practical synthesis method of the reagent PhenoFluor on decagram scale, provide a new formulation of PhenoFluor as a toluene solution, which should decrease challenges associated with the moisture sensitivity of the reagent, and expand the substrate scope of deoxyfluorination with PhenoFluor to heteroaromatics.