Merck
  • Silencing of H4R inhibits the production of IL-1β through SAPK/JNK signaling in human mast cells.
Silencing of H4R inhibits the production of IL-1β through SAPK/JNK signaling in human mast cells.
Journal of receptor and signal transduction research (2018-06-05)
Angel Jemima Ebenezer, Kavya Prasad, Sanjana Rajan, Elden Berla Thangam
ABSTRACT
Mast cell (MC) activation through H4R releases various inflammatory mediators which are associated with allergic asthma. To investigate the siRNA-mediated gene silencing effect of H4R on human mast cells (HMCs) functions and the activation of stress-activated protein kinases (SAPK)/jun amino-terminal kinases (JNK) signaling pathways for the release of ineterleukin-1β (IL-1β) in HMCs. H4R expression was analyzed by RT-PCR and western blotting in human mast cell line-1 (HMC-1) cells and H4RsiRNA transfected cells. The effect of H4RsiRNA and H4R-antagonist on H4R mediated MC functions such as intracellular Ca2+ release, degranulation, IL-6 and IL-1β release, and the activation SAPK/JNK signaling pathways were studied. HMC-1 cells were stimulated with 10 μM of histamine (His) and 4-methylhistamine (4-MH) and pretreated individually with H4R-antagonist JNJ7777120 (JNJ), histamine H1 receptor (H1R)-antagonist mepyramine, and signaling molecule inhibitors SP600125 (SP) and Bay117082. We found that the HMC-1 cells expressed H4R and H4RsiRNA treatment down regulated the H4R expression in HMC-1 cells. Both His and 4-MH induced the intracellular Ca2+ release and degranulation whereas; H4R siRNA and JNJ inhibited the effect. Furthermore, the activation of H4R caused the phosphorylation of SAPK/JNK pathways. H4R gene silencing and pretreatment with SP and JNJ decreased His and 4-MH induced phosphorylation of SAPK/JNK. We found that the activation of H4R caused the release of IL-1β (124.22 pg/ml) and IL-6 (122.50 pg/ml) on HMC-1 cells. Whereas, SAPK/JNK inhibitor (68.36 pg/ml) inhibited the H4R mediated IL-1β release. Taken together, the silencing of H4R inhibited the H4R mediated MC functions and SAPK/JNK phosphorylation. Furthermore, the H4R activation utilized SAPK/JNK signaling pathway for IL-1β release in HMC-1 cells.