Merck
  • Porcine Neonatal Pancreatic Cell Clusters Maintain Their Multipotency in Culture and After Transplantation.
Porcine Neonatal Pancreatic Cell Clusters Maintain Their Multipotency in Culture and After Transplantation.
Scientific reports (2018-05-31)
Wan-Chun Li, Chen-Yi Chen, Chen-Wei Kao, Pei-Chun Huang, Yi-Ta Hsieh, Tz-Yu Kuo, Tsai-Ying Chen, Hao-Yuan Chia, Jyuhn-Huarng Juang
ABSTRACT
Ductal epithelium is primarily detected in porcine neonatal pancreatic cell clusters (NPCCs) bearing grafts, suggesting that transplants might exhibit progenitor-like phenotypes. Here we found that soon after NPCC isolation, PDX1+/insulin- and SOX9+ pancreatic progenitor-like cells dramatically increased while dual-hormonal progenitor-like cells were routinely observed in NPCC culture. After transplantation (Tx), insulin+ cells increased and PDX1+ and SOX9+ cells gradually decreased in both non-diabetic (NDM) and streptozotocin-induced diabetic (DM) grafts over 2 months. Strikingly, a significantly higher percentage of insulin+ cells were detected in 9-day and 16-day, but not in 23-day, 30-day and 60-day grafts implying that hyperglycemia could only facilitate NPCC-derived β cells early post-Tx. A higher percentage of NPCC-derived β cells in early DM grafts was determined via an enhanced neogenic differentiation based on the detection of insulin+ cells budding out from PDX1+/SOX9+ epithelium. Interestingly, a drop in SOX9+ progenitor-like cells was detected 16 days post-Tx in DM grafts whilst PDX1+ cells do not show a significant difference until 60 days post-Tx between DM and NDM grafts, demonstrating that distinct progenitor-like populations fuel new β cells post-Tx. In conclusion, PDX1+/SOX9+ cells could be quickly activated after NPCC isolation, maintain their multipotency in culture and differentiate into new β cell post-Tx.