Merck
  • Comprehensive analysis of T cell leukemia signals reveals heterogeneity in the PI3 kinase-Akt pathway and limitations of PI3 kinase inhibitors as monotherapy.
Comprehensive analysis of T cell leukemia signals reveals heterogeneity in the PI3 kinase-Akt pathway and limitations of PI3 kinase inhibitors as monotherapy.
PloS one (2018-05-26)
Olga Ksionda, Marsilius Mues, Anica M Wandler, Lisa Donker, Milou Tenhagen, Jesse Jun, Gregory S Ducker, Ksenia Matlawska-Wasowska, Kevin Shannon, Kevan M Shokat, Jeroen P Roose
ABSTRACT
T cell acute lymphoblastic leukemia (T-ALL) is an aggressive hematologic cancer. Poly-chemotherapy with cytotoxic and genotoxic drugs causes substantial toxicity and more specific therapies targeting the underlying molecular lesions are highly desired. Perturbed Ras signaling is prevalent in T-ALL and occurs via oncogenic RAS mutations or through overexpression of the Ras activator RasGRP1 in ~65% of T-ALL patients. Effective small molecule inhibitors for either target do not currently exist. Genetic and biochemical evidence link phosphoinositide 3-kinase (PI3K) signals to T-ALL, PI3Ks are activated by Ras-dependent and Ras-independent mechanisms, and potent PI3K inhibitors exist. Here we performed comprehensive analyses of PI3K-Akt signaling in T-ALL with a focus on class I PI3K. We developed a multiplex, multiparameter flow cytometry platform with pan- and isoform-specific PI3K inhibitors. We find that pan-PI3K and PI3K γ-specific inhibitors effectively block basal and cytokine-induced PI3K-Akt signals. Despite such inhibition, GDC0941 (pan-PI3K) or AS-605240 (PI3Kγ-specific) as single agents did not efficiently induce death in T-ALL cell lines. Combination of GDC0941 with AS-605240, maximally targeting all p110 isoforms, exhibited potent synergistic activity for clonal T-ALL lines in vitro, which motivated us to perform preclinical trials in mice. In contrast to clonal T-ALL lines, we used a T-ALL cancer model that recapitulates the multi-step pathogenesis and inter- and intra-tumoral genetic heterogeneity, a hallmark of advanced human cancers. We found that the combination of GDC0941 with AS-605240 fails in such trials. Our results reveal that PI3K inhibitors are a promising avenue for molecular therapy in T-ALL, but predict the requirement for methods that can resolve biochemical signals in heterogeneous cell populations so that combination therapy can be designed in a rational manner.