Merck
  • Role of extracellular matrix in insulin-like growth factor (IGF) binding protein-2 regulation of IGF-II action in normal human osteoblasts.
Role of extracellular matrix in insulin-like growth factor (IGF) binding protein-2 regulation of IGF-II action in normal human osteoblasts.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society (2003-11-20)
Cheryl A Conover, Sundeep Khosla
ABSTRACT
Insulin-like growth factor binding protein-2 (IGFBP-2) in its native form had little affinity for extracellular matrix (ECM) derived from human or rat osteoblastic cells. However, in the presence of IGFs, IGFBP-2 binding to ECM was markedly enhanced, with IGF-II being more effective than IGF-I. IGF-II-enhanced binding of IGFBP-2 to ECM was specific for IGFBP-2 of the six known IGFBPs. In the presence of IGF-II, IGFBP-2 bound with high affinity to heparin-Sepharose, but not to type I collagen, fibronectin, or laminin. Furthermore, heparin and heparan sulfate, but not chondroitin sulfate, inhibited IGFBP-2/IGF-II binding to ECM. High salt (100 mM NaCl) inhibited, while CaCl(2) enhanced binding of IGFBP-2/IGF-II to ECM. In the presence of ECM, IGFBP-2/IGF-II was as effective as IGF-II alone in stimulating [3H]thymidine and [3H]proline incorporation and in inhibiting apoptosis in cultured human osteoblasts. On the other hand, IGFBP-2 was a potent inhibitor of IGF-II action in human breast and ovarian carcinoma cells. There was no difference between soluble and ECM-associated IGFBP-2 in affinity for IGF-I and IGF-II. These data suggest a unique mechanism for targeting an anabolic IGFBP-2/IGF-II complex in bone.