Merck
  • Regulation of human lymphocyte proliferation by a heterodimeric cytokine, IL-12 (cytotoxic lymphocyte maturation factor).
Regulation of human lymphocyte proliferation by a heterodimeric cytokine, IL-12 (cytotoxic lymphocyte maturation factor).
Journal of immunology (Baltimore, Md. : 1950) (1991-08-01)
M K Gately, B B Desai, A G Wolitzky, P M Quinn, C M Dwyer, F J Podlaski, P C Familletti, F Sinigaglia, R Chizonnite, U Gubler
ABSTRACT
IL-12 is a heterodimeric cytokine that was identified on the basis of its ability to synergize with IL-2 in the induction of cytotoxic effector cells and was originally called cytotoxic lymphocyte maturation factor (CLMF). IL-12 was also found to stimulate the proliferation of PHA-activated lymphoblasts which were greater than 90% CD3+ T cells. In this report we further characterize the effects of IL-12 on lymphocyte proliferation. Studies with purified subpopulations of PHA-activated lymphoblasts and with cloned lines of human T cells indicated that IL-12 caused the proliferation of activated T cells of both the CD4+ and CD8+ subsets. This effect of IL-12 was independent of IL-2 because it was not blocked by antibodies to either IL-2 or IL-2R. The maximum proliferation induced by IL-12 was 31 to 72% of the maximum caused by IL-2; however, IL-12 was active at a lower effective concentration (EC50 = 8.5 +/- 1.3 pM) than IL-2 (EC50 = 52 +/- 8 pM). Combination of suboptimal amounts of IL-12 and IL-2 resulted in additive proliferation, up to the maximum induced by IL-2 alone. IL-12 also caused the proliferation of lymphocytes activated by culture with IL-2 for 6 to 12 days. CD56+ NK cells were among the IL-12-responsive cells in the IL-2-activated lymphocyte population. Unlike IL-2 or IL-7, IL-12 caused little or no proliferation of resting peripheral blood mononuclear cells (PBMC). In this regard, IL-12 was similar to IL-4. However, IL-12 could enhance the proliferation of resting PBMC caused by suboptimal amounts of IL-2, whereas IL-4 inhibited IL-2-induced PBMC proliferation. Thus, IL-12 is a growth factor for activated human T cells and NK cells; however, its spectrum of lymphocyte growth-promoting properties is distinct from that of IL-2, IL-4, or IL-7.