Merck
  • Expression of the Alpha8 Integrin Chain Facilitates Phagocytosis by Renal Mesangial Cells.
Expression of the Alpha8 Integrin Chain Facilitates Phagocytosis by Renal Mesangial Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-03-16)
Ines Marek, Robert Becker, Fabian B Fahlbusch, Carlos Menendez-Castro, Wolfgang Rascher, Christoph Daniel, Gudrun Volkert, Andrea Hartner
ABSTRACT
Healing of mesangioproliferative glomerulonephritis involves degradation of excess extracellular matrix, resolution of hypercellularity by apoptosis and phagocytosis of apoptotic cells. Integrin receptors participate in the regulation of phagocytosis. In mice deficient for alpha8 integrin (Itga8-/-) healing of glomerulonephritis is delayed. As Itga8 is abundant in mesangial cells (MC) which are non-professional phagocytes, we hypothesized that Itga8 facilitates phagocytosis of apoptotic cells and matrix components by MC. MC were isolated from wild type (WT) and Itga8-/- mice. Latex beads were coated with matrix components. Apoptosis was induced by cisplatin in macrophages and in DiI-stained MC. After coincubation of latex beads or apoptotic cells with MC, the phagocytosis rate was detected in WT and Itga8-/- MC via fluorescence microscopy and FACS analysis. Itga8-/- MC showed reduced phagocytosis of matrix-coated beads and apoptotic cells compared to WT MC. Reduction of stress fibers was observed in Itga8-/- compared to WT MC. Inhibition of cytoskeletal reorganization by inhibition of Rac1 or ROCK during phagocytosis significantly decreased the rate of phagocytosis by WT MC but not by Itga8-/- MC. The expression of Itga8 facilitates phagocytosis in MC, likely mediated by Itga8-cytoskeleton interactions. An impairment of MC phagocytosis might thus contribute to a delayed glomerular regeneration in Itga8-/- mice.