Merck
  • A Facile Approach for Synthesis and Intracellular Delivery of Size Tunable Cationic Peptide Functionalized Gold Nanohybrids in Cancer Cells.
A Facile Approach for Synthesis and Intracellular Delivery of Size Tunable Cationic Peptide Functionalized Gold Nanohybrids in Cancer Cells.
Bioconjugate chemistry (2018-03-01)
Kavita Bansal, Mohammad Aqdas, Munish Kumar, Rajni Bala, Sanpreet Singh, Javed N Agrewala, O P Katare, Rohit K Sharma, Nishima Wangoo
ABSTRACT
Peptide-based drug delivery systems have become a mainstay in the contemporary medicinal field, resulting in the design and development of better pharmaceutical formulations. However, most of the available reports employ tedious multiple reaction steps for the conjugation of bioactive cationic peptides with drug delivery vehicles. To overcome these limitations, the present work describes a one-step approach for facile and time efficient synthesis of highly cationic cell penetrating peptide functionalized gold nanoparticles and their intracellular delivery. The nanoconstruct was synthesized by the reduction of gold metal ions utilizing cell penetrating peptide (CPP), which facilitated the simultaneous synthesis of metal nanoparticles and the capping of the peptide over the nanoparticle surface. The developed nanoconstruct was thoroughly characterized and tested for intracellular delivery into HeLa cells. Intriguingly, a high payload of cationic peptide over gold particles was achieved, in comparison to conventional conjugation methods. Moreover, this method also provides the ability to control the size and peptide payload of nanoparticles. The nanoconstructs produced showed enhanced cancer cell penetration (μM) and significant cytotoxic effect compared to unlabeled gold nanoparticles. Therefore, this novel approach may also have significant future potential to kill intracellular hidden dreaded pathogens like the human immunodeficiency virus, Mycobacterium tuberculosis, and so forth.
Sigma-Aldrich
Fmoc-Lys(Boc)-OH, ≥98.0% (HPLC)