Merck
  • Bafilomycin A1 increases the sensitivity of tongue squamous cell carcinoma cells to cisplatin by inhibiting the lysosomal uptake of platinum ions but not autophagy.
Bafilomycin A1 increases the sensitivity of tongue squamous cell carcinoma cells to cisplatin by inhibiting the lysosomal uptake of platinum ions but not autophagy.
Cancer letters (2018-03-11)
Hong-Ying Chu, Wei Wang, Xue Chen, Yu-E Jiang, Rui Cheng, Xiao Qi, Zhao-Ming Zhong, Mu-Sheng Zeng, Xiao-Feng Zhu, Chuan-Zheng Sun
ABSTRACT
The role of autophagy in tongue squamous cell carcinoma (TSCC) cisplatin resistance is unclear. We aimed to identify a possible synergistic effect of autophagy inhibitors and cisplatin in TSCC cells and explore the underlying mechanism. Our results indicate that autophagic flux was high in TSCC cells; Autophagy inhibitor bafilomycin A1 increased cisplatin cytotoxicity in TSCC cells by inhibiting lysosomal uptake of platinum and enhancing intracellular platinum ion binding to DNA; Autophagy gene (Atg5) knockout in TSCC cells did not duplicate the above-mentioned sensitization of bafilomycin A1. Furthermore, we found that cisplatin resistance of TSCC cells was related to cisplatin inducing lysosome biogenesis in a TFEB-dependent manner, which was regulated by c-Abl. In summary, this is the first study to show that Bafilomycin A1 increases the sensitivity of TSCC cells to cisplatin by inhibiting lysosomal function but not autophagy. Lysosomes may be a potential target to increase cisplatin cytotoxicity toward TSCC cells.