Merck
  • Mtf2-PRC2 control of canonical Wnt signaling is required for definitive erythropoiesis.
Mtf2-PRC2 control of canonical Wnt signaling is required for definitive erythropoiesis.
Cell discovery (2018-05-08)
Janet L Manias Rothberg, Harinad B Maganti, Hani Jrade, Christopher J Porter, Gareth A Palidwor, Christopher Cafariello, Hannah L Battaion, Safwat T Khan, Theodore J Perkins, Robert F Paulson, Caryn Y Ito, William L Stanford
ABSTRACT
Polycomb repressive complex 2 (PRC2) accessory proteins play substoichiometric, tissue-specific roles to recruit PRC2 to specific genomic loci or increase enzymatic activity, while PRC2 core proteins are required for complex stability and global levels of trimethylation of histone 3 at lysine 27 (H3K27me3). Here, we demonstrate a role for the classical PRC2 accessory protein Mtf2/Pcl2 in the hematopoietic system that is more akin to that of a core PRC2 protein. Mtf2 -/- erythroid progenitors demonstrate markedly decreased core PRC2 protein levels and a global loss of H3K27me3 at promoter-proximal regions. The resulting de-repression of transcriptional and signaling networks blocks definitive erythroid development, culminating in Mtf2 -/- embryos dying by e15.5 due to severe anemia. Gene regulatory network (GRN) analysis demonstrated Mtf2 directly regulates Wnt signaling in erythroblasts, leading to activated canonical Wnt signaling in Mtf2-deficient erythroblasts, while chemical inhibition of canonical Wnt signaling rescued Mtf2-deficient erythroblast differentiation in vitro. Using a combination of in vitro, in vivo and systems analyses, we demonstrate that Mtf2 is a critical epigenetic regulator of Wnt signaling during erythropoiesis and recast the role of polycomb accessory proteins in a tissue-specific context.
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %