- Harnessing Alkyl Amines as Electrophiles for Nickel-Catalyzed Cross Couplings via C-N Bond Activation.
Harnessing Alkyl Amines as Electrophiles for Nickel-Catalyzed Cross Couplings via C-N Bond Activation.
Journal of the American Chemical Society (2017-04-01)
Corey H Basch, Jennie Liao, Jianyu Xu, Jacob J Piane, Mary P Watson
PMID28359153
ABSTRACT
We developed a strategy to harness alkyl amines as alkylating agents via C-N bond activation. This Suzuki-Miyaura cross coupling of alkylpyridinium salts, readily formed from primary amines, is the first example of a metal-catalyzed cross coupling via C-N bond activation of an amine with an unactivated alkyl group. This reaction enjoys broad scope and functional group tolerance. Primary and secondary alkyl groups can be installed. Preliminary studies suggest a NiI/NiIII catalytic cycle.