Merck
Aprepitant and fosaprepitant drug interactions: a systematic review.
British journal of clinical pharmacology (2017-05-05)
Priya Patel, J Steven Leeder, Micheline Piquette-Miller, L Lee Dupuis
ABSTRACT
Aprepitant and fosaprepitant, commonly used for the prevention of chemotherapy-induced nausea and vomiting, alter cytochrome P450 activity. This systematic review evaluates clinically significant pharmacokinetic drug interactions with aprepitant and fosaprepitant and describes adverse events ascribed to drug interactions with aprepitant or fosaprepitant. We systematically reviewed the literature to September 11, 2016, to identify articles evaluating drug interactions involving aprepitant/fosaprepitant. The clinical significance of each reported pharmacokinetic drug interaction was evaluated based on the United States Food and Drug Administration guidance document on conducting drug interaction studies. The probability of an adverse event reported in case reports being due to a drug interaction with aprepitant/fosaprepitant was determined using the Drug Interaction Probability Scale. A total of 4377 publications were identified. Of these, 64 met inclusion eligibility criteria: 34 described pharmacokinetic drug interactions and 30 described adverse events ascribed to a drug interaction. Clinically significant pharmacokinetic interactions between aprepitant/fosaprepitant and bosutinib PO, cabazitaxel IV, cyclophosphamide IV, dexamethasone PO, methylprednisolone IV, midazolam PO/IV, oxycodone PO and tolbutamide PO were identified, as were adverse events resulting from an interaction between aprepitant/fosaprepitant and alcohol, anthracyclines, ifosfamide, oxycodone, quetiapine, selective serotonin reuptake inhibitors/serotonin-norepinephrine reuptake inhibitors and warfarin. The potential for a drug interaction with aprepitant and fosaprepitant should be considered when selecting antiemetic therapy.