Merck
  • Brazilin exerts protective effects against renal ischemia-reperfusion injury by inhibiting the NF-κB signaling pathway.
Brazilin exerts protective effects against renal ischemia-reperfusion injury by inhibiting the NF-κB signaling pathway.
International journal of molecular medicine (2016-06-02)
Yanyan Jia, Jinyi Zhao, Meiyou Liu, Bingling Li, Ying Song, Yuwen Li, Aidong Wen, Lei Shi
ABSTRACT
Renal ischemia-reperfusion (I/R) injury is associated with high morbidity and mortality as there is currently no available effective therapeutic strategy with which to treat this injury. Thus, the aim of this study was to investigate the potential protective effects of brazilin, a major active component of the Chinese medicine Caesalpinia sappan L., against renal I/R injury in vitro and in vivo. Rats were subjected to removal of the right kidney and I/R injury to the left kidney (ischemia for 45 min followed by reperfusion for 24 h). Treatment with brazilin (30 mg/kg, administered intravenously at 30 min prior to ischemia) led to the reversal of I/R-induced changes in serum creatinine (Scr) and blood urea nitrogen (BUN) levels, and also attenuated the histopathological damage induced by I/R. Furthermore, TUNEL assay revealed that brazilin reduced cell necrosis, and significantly decreased the expression of tumor necrosis factor (TNF)-α and interleukin (IL)-1β in renal tissue. Moreover, HK-2 cells were used in order to elucidate the mechanisms responsible for the protective effects of brazilin. The levels of phosphorylated IκBα and the nuclear translocation of nuclear factor-κB (NF-κB) were all evidently decreased by brazilin. These findings suggested that pre-treatment with brazilin protects against I/R-induced renal damage and suppresses the inflammatory response by inhibiting the activation of the NF-κB signaling pathway.