Merck
  • Alternative Lengthening of Telomeres and Loss of DAXX/ATRX Expression Predicts Metastatic Disease and Poor Survival in Patients with Pancreatic Neuroendocrine Tumors.
Alternative Lengthening of Telomeres and Loss of DAXX/ATRX Expression Predicts Metastatic Disease and Poor Survival in Patients with Pancreatic Neuroendocrine Tumors.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-07-14)
Aatur D Singhi, Ta-Chiang Liu, Justin L Roncaioli, Dengfeng Cao, Herbert J Zeh, Amer H Zureikat, Allan Tsung, J Wallis Marsh, Kenneth K Lee, Melissa E Hogg, Nathan Bahary, Randall E Brand, Kevin M McGrath, Adam Slivka, Kristi L Cressman, Kimberly Fuhrer, Roderick J O'Sullivan
ABSTRACT
Pancreatic neuroendocrine tumors (PanNET) are a heterogeneous group of neoplasms with increasing incidence and unpredictable behavior. Whole-exome sequencing has identified recurrent mutations in the genes DAXX and ATRX, which correlate with loss of protein expression and alternative lengthening of telomeres (ALT). Both ALT and DAXX/ATRX loss were initially reported to be associated with a favorable prognosis; however, recent studies suggest the contrary. Our aims were to assess the prevalence and prognostic significance of ALT and DAXX/ATRX in both primary and metastatic PanNETs. Telomere-specific FISH and DAXX/ATRX IHC was performed on a multi-institutional cohort of 321 patients with resected PanNET and 191 distant metastases from 52 patients. These results were correlated with clinicopathologic features, including disease-free survival (DFS) and disease-specific survival (DSS). The prevalence of ALT and DAXX/ATRX loss in resected PanNETs was 31% and 26%, respectively, and associated with larger tumor size, higher WHO grade, lymph node metastasis, and distant metastasis (P < 0.001). The 5-year DFS and 10-year DSS of patients with ALT-positive and DAXX/ATRX-negative PanNETs were 40% and 50%, respectively, as compared with 96% and 89%, respectively, for wild-type PanNETs. Among distant metastases, ALT and DAXX/ATRX loss was 67% and 52%, respectively, and only occurred in the setting of an ALT-positive and DAXX/ATRX-negative primary PanNET. By multivariate analysis, both ALT and DAXX/ATRX loss were negative, independent prognostic factors for DFS. ALT and DAXX/ATRX loss in PanNETs was associated with shorter DFS and DSS and likely plays a significant role in driving metastatic disease. Clin Cancer Res; 23(2); 600-9. ©2016 AACR.