Merck
  • APC sets the Wnt tone necessary for cerebral cortical progenitor development.
APC sets the Wnt tone necessary for cerebral cortical progenitor development.
Genes & development (2017-09-17)
Naoki Nakagawa, Jingjun Li, Keiko Yabuno-Nakagawa, Tae-Yeon Eom, Martis Cowles, Tavien Mapp, Robin Taylor, E S Anton
ABSTRACT
Adenomatous polyposis coli (APC) regulates the activity of β-catenin, an integral component of Wnt signaling. However, the selective role of the APC-β-catenin pathway in cerebral cortical development is unknown. Here we genetically dissected the relative contributions of APC-regulated β-catenin signaling in cortical progenitor development, a necessary early step in cerebral cortical formation. Radial progenitor-specific inactivation of the APC-β-catenin pathway indicates that the maintenance of appropriate β-catenin-mediated Wnt tone is necessary for the orderly differentiation of cortical progenitors and the resultant formation of the cerebral cortex. APC deletion deregulates β-catenin, leads to high Wnt tone, and disrupts Notch1 signaling and primary cilium maintenance necessary for radial progenitor functions. β-Catenin deregulation directly disrupts cilium maintenance and signaling via Tulp3, essential for intraflagellar transport of ciliary signaling receptors. Surprisingly, deletion of β-catenin or inhibition of β-catenin activity in APC-null progenitors rescues the APC-null phenotype. These results reveal that APC-regulated β-catenin activity in cortical progenitors sets the appropriate Wnt tone necessary for normal cerebral cortical development.
Sigma-Aldrich
Anti-Tbr1 Antibody, from chicken, purified by affinity chromatography