Merck
  • Ramelteon Improves Post-traumatic Stress Disorder-Like Behaviors Exhibited by Fatty Acid-Binding Protein 3 Null Mice.
Ramelteon Improves Post-traumatic Stress Disorder-Like Behaviors Exhibited by Fatty Acid-Binding Protein 3 Null Mice.
Molecular neurobiology (2017-05-19)
Yasushi Yabuki, Ibuki Takahata, Kazuya Matsuo, Yuji Owada, Kohji Fukunaga
ABSTRACT
We previously reported that fatty acid-binding protein 3 (FABP3) knockout (Fabp3