Merck
  • Impaired generation of hepatitis B virus-specific memory B cells in HIV infected individuals following vaccination.
Impaired generation of hepatitis B virus-specific memory B cells in HIV infected individuals following vaccination.
Vaccine (2010-04-02)
Nishaki Mehta, Coleen K Cunningham, Patricia Flynn, Joyce Pepe, Stephen Obaro, Bill G Kapogiannis, James Bethel, Katherine Luzuriaga
ABSTRACT
Hepatitis B-specific memory B cell (HSMBC) frequencies were measured following hepatitis B vaccination in 15 HIV uninfected and 12 HIV infected adolescents. HSMBC were detected at significantly lower frequencies in HIV infected than in HIV uninfected individuals. The detection of HBsAb >10mIU/ml at study week 28 was strongly associated with the detection of HSMBC and a direct correlation between HBsAb titers and HSMBC frequencies was observed. In HIV uninfected individuals, antibody titers >1000mIU/ml were associated with higher HSMC frequencies. Lower HSMBC frequencies, reduced memory B cell (MBC) proliferation, and altered B cell phenotypes were measured in viremic HIV infected individuals compared with aviremic HIV infected or HIV uninfected individuals.