Skip to Content
Merck
Undecylenic acid ≥96%, FG

Undecylenic acid

Undecylenic acid ≥96%, FG
Synonym(s):
10-Undecenoic acid
Linear Formula:
CH2=CH(CH2)8COOH
CAS Number:
Molecular Weight:
184.28
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing