Tetraoctylphosphonium bromide
Synonym(s):
Tetraoctylphosphonium bromide
Empirical Formula (Hill Notation):
C32H68BrP
CAS Number:
Molecular Weight:
563.76
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing