
Sodium D-isoascorbate monohydrate

Synonym(s):
D-Isoascorbic acid sodium salt monohydrate, Sodium erythorbate monohydrate
Empirical Formula (Hill Notation):
C6H7NaO6 · H2O
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing