![Safranine O (C.I. 50240) for microscopy Certistain®](/deepweb/assets/sigmaaldrich/product/images/306/432/b9468f56-d73b-4992-ad4d-43214453654d/640/b9468f56-d73b-4992-ad4d-43214453654d.jpg)
Safranine O (C.I. 50240)
![Safranine O (C.I. 50240) for microscopy Certistain®](/deepweb/assets/sigmaaldrich/product/images/306/432/b9468f56-d73b-4992-ad4d-43214453654d/640/b9468f56-d73b-4992-ad4d-43214453654d.jpg)
Synonym(s):
Safranine O (C.I. 50240)
Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing