
Potassium hydrogen diiodate

Synonym(s):
Potassium hydrogen diiodate
Linear Formula:
KH(IO3)2
CAS Number:
Molecular Weight:
389.91
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing