
Poncirin

Synonym(s):
(S)-5,7-Dihydroxy-4′-methoxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], 4′-O-Methylnaringin, Isosakuranetin-7-neohesperidoside
Empirical Formula (Hill Notation):
C28H34O14
CAS Number:
Molecular Weight:
594.56
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1