
Phloxin B (C.I. 45410)

Synonym(s):
Phloxin B (C.I. 45410)
Empirical Formula (Hill Notation):
C20H2Br4Cl4Na2O5
CAS Number:
Molecular Weight:
829.63
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing