Skip to Content
Merck
Pentadecanoic acid analytical standard

Pentadecanoic acid

Pentadecanoic acid analytical standard
Synonym(s):
Pentadecanoic acid, C15:0, NSC 28486, Pentadecylic acid
Linear Formula:
CH3(CH2)13COOH
CAS Number:
Molecular Weight:
242.40
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing