Skip to Content
Merck
Methyl p-anisate ≥99%, FG

Methyl p-anisate

Methyl p-anisate ≥99%, FG
Synonym(s):
p-Anisic acid methyl ester, Methyl 4-methoxybenzoate
Linear Formula:
CH3OC6H4CO2CH3
CAS Number:
Molecular Weight:
166.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing