![Methyl myristoleate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/304/892/5dceb139-559b-4067-be87-87afcb8d45a4/640/5dceb139-559b-4067-be87-87afcb8d45a4.png)
Methyl myristoleate
![Methyl myristoleate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/304/892/5dceb139-559b-4067-be87-87afcb8d45a4/640/5dceb139-559b-4067-be87-87afcb8d45a4.png)
Synonym(s):
Methyl cis-9-tetradecenoate, Myristoleic acid methyl ester
Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
Products
Product Category
Brand
Formula Weight
Purity
Format
Quality Segment
Functional Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing