Skip to Content
Merck
Lead nitrate NIST® SRM® 928

Lead nitrate

Lead nitrate NIST® SRM® 928
Synonym(s):
Lead(II) nitrate, Lead dinitrate
Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21

Products

Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing