
Ipratropium bromide Related Compound B

Synonym(s):
(1R,3r,5S,8s)-3-[[(2RS)-3-Hydroxy-2-phenylpropanoyl]oxy]-8-methyl-8-(1-methylethyl)-8-azoniabicyclo[3.2.1]octane bromide
Empirical Formula (Hill Notation):
C20H30BrNO3
CAS Number:
Molecular Weight:
412.36
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing